Sodium hydroxide 20% (w/v) in aqueous solution (200 g/L)

Supplier: Ricca Chemical

7270-5
RC7270-5EA 1053.86 CAD
RC7270-5
Sodium hydroxide 20% (w/v) in aqueous solution (200 g/L)
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Assay 20 ± 0.04 % (w/v)
NIST SRM 723


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR