Sodium azide 5% (w/v) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Sodium triazide
7144.8-16
RC7144.8-16EA
142.63
CAD
RC7144.8-16
Sodium azide 5% (w/v) in aqueous solution
Sodium azide
Formula:
NaN₃ MW: 65.01 g/mol |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 UN: 1687 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...