Picric Acid-Acetone for Brown & Brinn

Supplier: Ricca Chemical

R5861100-4A R5861100-500A
10036-326EA 235.42 CAD
10036-326 10036-328
Picric Acid-Acetone for Brown & Brinn
Picric Acid-Acetone for Brown & Brinn

Acetone 67-64-1 ACS (99.87%)
Picric Acid 88-89-1 ACS (0.09%)
Water 7732-18-5 ACS (0.04%)
Test Specification Result : Appearance Yellow liquid
Volumetric glassware complies with Class A tolerance requirements of ASTM E 288 and NIST Circular 434; it is calibrated before first use and
recalibrated regularly in accordance with ASTM E 542 and NIST Procedure NBSIR 74-461. Balances are calibrated regularly with weights certified
traceable to the NIST national mass standard. Thermometers and temperature probes are calibrated before first use and recalibrated regularly with a thermometer traceable to NIST standards. All products are prepared according to master documents that assure manufacture according to validatedmethods. Batch records document raw material traceability and production and testing history for each lot manufactured.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR