Orthophosphoric acid 25% (v/v) in aqueous solution (1 + 3)
Supplier: Ricca Chemical
|
Danger
|
R5851000-1A
R5851000-20F
RC585132EA
82.16
CAD
RC585132
RCR5851000-20F
Orthophosphoric acid 25% (v/v) in aqueous solution (1 + 3)
Orthophosphoric acid
Formula:
H₃PO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 UN: 1805 ADR: 8,III |
Specification Test Results
Appearance | Colorless to light yellow liquid |
Assay (vs. Sodium Hydroxide/Thymolphthalein) | 24.96-25.04 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...