Methanol absolute ACS, Reagent Grade for dilution
Supplier: Ricca Chemical
Synonyms:
Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
4829-1
4829-32
RC4829-1EA
447.14
CAD
RC4829-1
RC4829-32
Methanol absolute ACS, Reagent Grade for dilution
Methanol
Formula:
CH₃OH MW: 32.04 g/mol Boiling Pt: 64.6 Melting Pt: -98 Flash Pt: 11 |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 UN: 1230 ADR: 3 II Merck Index: 13,05984 |
|
Specification Test Results
Residue After Evaporation | 0.00001 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...