Keller's reagent etchant solution
Supplier: Ricca Chemical
|
Danger
|
R4250000-100A
R4250000-1A
RCR4250000100AEA
483.48
CAD
RCR4250000100A
RCR4250000-1A
Keller's reagent etchant solution
Keller's reagent
Storage Temperature:
Ambient |
UN:
3264 ADR: 8,III |
Specification Test Results
Appearance | Colorless liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...