2-Propanol ≥70% (v/v) in aqueous solution
Supplier: Ricca Chemical
4210-16
4210-1G
4210-2.5
4210-32
RC4210-16EA
82.16
CAD
RC4210-16
RC4210-1G
RC4210-2.5
RC4210-32
2-Propanol ≥70% (v/v) in aqueous solution
2-Propanol
Formula:
(CH₃)₂CHOH |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Specification Test Results
Specific Gravity | 70 ± 2 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...