Citric acid 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acidHydroxytricarballylic acid

2120-32
RC2120-32EA 116.24 CAD
RC2120-32
Citric acid 10% (w/v) in aqueous solution
Citric acid

Manufactured from USP grade material.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0405 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
https://ca.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg

Order Now

Specification Test Results

Assay 10 ± 0.03 % (w/v)
NIST SRM 84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR