Ceric ammonium sulfate 0.100 N (N/10) in 1 N sulphuric Acid

Supplier: Ricca Chemical
Danger

1890-1 1890-32
RC1890-1EA 398.16 CAD
RC1890-1 RC1890-32
Ceric ammonium sulfate 0.100 N (N/10) in 1 N sulphuric Acid
Ceric ammonium sulfate
Formula: H₈CeN₂O₁₂S₃
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00148851
CAS Number: 13840-04-5

Order Now

Specification Test Results

Assay 0.1 ± 0.0001 N at 20 °C
NIST SRM 83


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR