Acetic acid 56% (w/w) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

140.5-1 140.5-16 140.5-32
RC140.5-1EA 364.98 CAD
RC140.5-1 RC140.5-16 RC140.5-32
Acetic acid 56% (w/w) in aqueous solution
Acetic acid
Formula: H₃CCOOH
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 56 ± 0.06 % (w/w)
NIST SRM 84


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR