Transfection Reagent, 293-FREE, Novagen, MilliporeSigma
Supplier: MilliporeSigma
72181-3 72181-4 72181-5
CA80018-034EA 1185.45 CAD
CA80018-034 CA80018-036 CA80018-038
Transfection Reagent, 293-FREE, Novagen, MilliporeSigma
Transfection Reagents
Has a unique polycationic liposomal formulation and is designed expressly for transfection of HEK 293 cells grown in suspension culture
Derived from non-animal sources. Provided as a sterile, ready-to use solution.