Hei-VAP Industrial Safety Rotary Evaporators, 220 V
Supplier: Heidolph NA, LLC
036072005CA 036072015CA 036072020CA 036072025CA 036072040CA 036072000CA 036072010CA
CA10018-636EA 140027.75 CAD
CA10018-636 CA10018-638 CA10018-640 CA10018-648 CA10018-654 CA10018-656 CA10018-682 CA10018-684
Hei-VAP Industrial Safety Rotary Evaporators, 220 V
Evaporators Rotary Evaporators
Hei-VAP Industrial Safety Rotary Evaporators offer the highest safety and integrated vacuum control.
  • 220 V model
  • Excellent user safety with high-impact transparent PMMA door
  • Non-fogging safety glass and metal frame guard hood provides excellent user protection
  • The evaporation flask is illuminated during operation for increased visibility
  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
Evaporators come standard with integrated refill water system, spillover preventions, and a release valve on the bottom. Optional base cart is fully maneuverable with removable panels for storage of vacuum pump, glassware, solvents, or accessories.
Certifications: Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ).
Ordering information: Glass set included. Base and all other accessories not included.
Caution: 220 V model.