Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
|
Management of Change (MOC) category = N
9387-03
9387-01
9387-01
BAKR9387-03
JT9387-1EA
311.43
CAD
JT9387-3
JT9387-1
CAJT9387-1
CAJT9387-3
Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 UN: 1790 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Color (APHA) | ≤10 |
Assay (HF)(w/w) | 47 - 51 % |
Chloride (Cl) | ≤4 ppm |
Fluosilicic Acid (SiF₆) | ≤20 ppm |
Total Phosphorus (P) | ≤0.05 ppm |
Total Sulphur (S) | ≤0.1 ppm |
Trace Impurities - Aluminum (Al) | ≤1 ppb |
Trace Impurities - Antimony (Sb) | ≤0.2 ppb |
Trace Impurities - Arsenic (As) | ≤0.5 ppb |
Trace Impurities - Barium (Ba) | ≤0.1 ppb |
Trace Impurities - Beryllium (Be) | ≤0.1 ppb |
Trace Impurities - Bismuth (Bi) | ≤0.1 ppb |
Trace Impurities - Boron (B) | ≤1 ppb |
Trace Impurities - Cadmium (Cd) | ≤0.1 ppb |
Trace Impurities - Calcium (Ca) | ≤1 ppb |
Trace Impurities - Cerium (Ce) | ≤0.1 ppb |
Trace Impurities - Cesium (Cs) | ≤0.1 ppb |
Trace Impurities - Chromium (Cr) | ≤1 ppb |
Trace Impurities - Cobalt (Co) | ≤0.1 ppb |
Trace Impurities - Copper (Cu) | ≤0.5 ppb |
Trace Impurities - Dysprosium (Dy) | ≤0.1 ppb |
Trace Impurities - Erbium (Er) | ≤0.1 ppb |
Trace Impurities - Europium (Eu) | ≤0.1 ppb |
Trace Impurities - Gadolinium (Gd) | ≤0.1 ppb |
Trace Impurities - Gallium (Ga) | ≤0.1 ppb |
Trace Impurities - Germanium (Ge) | ≤0.1 ppb |
Trace Impurities - Gold (Au) | ≤0.2 ppb |
Trace Impurities - Hafnium (Hf) | ≤0.1 ppb |
Trace Impurities - Holmium (Ho) | ≤0.1 ppb |
Trace Impurities - Indium (In) | ≤0.1 ppb |
Trace Impurities - Iron (Fe) | ≤1 ppb |
Trace Impurities - Lanthanum (La) | ≤0.1 ppb |
Trace Impurities - Lead (Pb) | ≤0.1 ppb |
Trace Impurities - Lithium (Li) | ≤0.1 ppb |
Trace Impurities - Lutetium (Lu) | ≤0.1 ppb |
Trace Impurities - Magnesium (Mg) | ≤1 ppb |
Trace Impurities - Manganese (Mn) | ≤0.1 ppb |
Trace Impurities - Mercury (Hg) | ≤1 ppb |
Trace Impurities - Molybdenum (Mo) | ≤0.1 ppb |
Trace Impurities - Neodymium (Nd) | ≤0.1 ppb |
Trace Impurities - Nickel (Ni) | ≤0.5 ppb |
Trace Impurities - Niobium (Nb) | ≤0.1 ppb |
Trace Impurities - Palladium (Pd) | |
Trace Impurities - Platinum (Pt) | |
Trace Impurities - Potassium (K) | ≤1 ppb |
Trace Impurities - Praseodymium (Pr) | ≤0.1 ppb |
Trace Impurities - Rhenium (Re) | ≤0.1 ppb |
Trace Impurities - Rhodium (Rh) | ≤0.1 ppb |
Trace Impurities - Rubidium (Rb) | ≤0.1 ppb |
Trace Impurities - Ruthenium (Ru) | ≤0.1 ppb |
Trace Impurities - Samarium (Sm) | ≤0.1 ppb |
Trace Impurities - Scandium (Sc) | ≤0.1 ppb |
Trace Impurities - Selenium (Se) | ≤1 ppb |
Trace Impurities - Silver (Ag) | ≤0.5 ppb |
Trace Impurities - Sodium (Na) | ≤1 ppb |
Trace Impurities - Strontium (Sr) | ≤0.1 ppb |
Trace Impurities - Tantalum (Ta) | |
Trace Impurities - Tellurium (Te) | ≤0.1 ppb |
Trace Impurities - Terbium (Tb) | ≤0.1 ppb |
Trace Impurities - Thallium (Tl) | ≤0.1 ppb |
Trace Impurities - Thorium (Th) | ≤0.1 ppb |
Trace Impurities - Thulium (Tm) | ≤0.1 ppb |
Trace Impurities - Tin (Sn) | ≤0.5 ppb |
Trace Impurities - Titanium (Ti) | ≤1 ppb |
Trace Impurities - Tungsten (W) | ≤0.5 ppb |
Trace Impurities - Uranium (U) | ≤0.1 ppb |
Trace Impurities - Vanadium (V) | ≤0.1 ppb |
Trace Impurities - Ytterbium (Yb) | ≤0.1 ppb |
Trace Impurities - Yttrium (Y) | ≤0.1 ppb |
Trace Impurities - Zinc (Zn) | ≤1 ppb |
Trace Impurities - Zirconium (Zr) | ≤0.1 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...