Reagent alcohol 70% (v/v) (denatured with methanol and isopropanol)

Supplier: Ricca Chemical
Danger

Synonyms: Reagent alcohol

R2546700-4A 2546.70-5 R2546700-3.8A 2546.70-32 2546.70-2.5 2546.70-1SC
RCR2546700-3.8AEA 225.72 CAD
RC2546.70-2.5 RC2546.70-1 RC2546.70-32 RCR2546700-3.8A RC2546.70-5 RC2546.70-1SC
Reagent alcohol 70% (v/v) (denatured with methanol and isopropanol)
Alcohol

Reagent Alcohol (Denatured Alcohol), General use cleaning solutions and solvents.

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR