Anti-PTPRC Rat Antibody (redFluor® 710) [clone: 30-F11]

Supplier: Tonbo Biosciences
80-0451-U025 80-0451-U100
10141-102EA 94.8 CAD
10141-102 10141-104
Anti-PTPRC Rat Antibody (redFluor® 710) [clone: 30-F11]
Antibodies
The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB and CD45RO.

The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.

Type: Primary
Antigen: PTPRC
Clonality:
Clone: 30-F11
Conjugation: redFluor® 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR