Protector® Radioisotope Laboratory Hoods, Labconco®
Supplier: Labconco
120400000 120400001 120400002 120400020 120400021 120410000 120410001 120410002 120410020 120410021 120500000 120500001 120500002 120500020 120500021 120510000 120510001 120510002 120510020 120510021 120600000 120600001 120600002 120600020 120600021 120610000 120610001 120610002 120610020 120610021 120800000 120800001 120800002 120800020 120800021 120810000 120810001 120810002 120810020 120810021
89523-348EA 35794.9 CAD
89523-348 89523-350 89523-352 89523-354 89523-356 89523-358 89523-360 89523-362 89523-364 89523-366 89523-368 89523-370 89523-372 89523-374 89523-376 89523-378 89523-380 89523-382 89523-384 89523-386 89523-388 89523-390 89523-392 89523-394 89523-396 89523-398 89523-400 89523-402 89523-404 89523-406 89523-408 89523-410 89523-412 89523-414 89523-416 89523-418 89523-420 89523-422 89523-424 89523-426
Protector® Radioisotope Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Designed to give maximum protection for radiochemical applications.
Components for converting either or both fixtures to vacuum and cold water are provided. Hood is factory prepared for up to eight service fixtures and four electrical duplexes. T8 fluorescent lights with vapor-proof covers provide a bright, nonglare working area.
Stainless steel interior.
One-piece, seamless, Type 304 stainless steel liner has radiused corners and integral bottom. Supporting work surface is included. Free of joints, cracks, and gaskets to minimize contamination build-up. Exterior is glacier whiteMore Product Information