Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco®
Supplier: Labconco
130400000 130400001 130400002 130400020 130400021 130410000 130410001 130410002 130410020 130410021 130500000 130500001 130500002 130500020 130500021 130510000 130510001 130510002 130510020 130510021 130600000 130600001 130600002 130600020 130600021 130610000 130610001 130610002 130610020 130610021 130800000 130800001 130800002 130800020 130800021 130810000 130810001 130810002 130810020 130810021
89523-268EA 38868 CAD
89523-268 89523-270 89523-272 89523-274 89523-276 89523-278 89523-280 89523-282 89523-284 89523-286 89523-288 89523-290 89523-292 89523-294 89523-296 89523-298 89523-300 89523-302 89523-304 89523-306 89523-308 89523-310 89523-312 89523-314 89523-316 89523-318 89523-320 89523-322 89523-324 89523-326 89523-328 89523-330 89523-332 89523-334 89523-336 89523-338 89523-340 89523-342 89523-344 89523-346
Protector® Stainless Steel Perchloric Acid Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Interior is one piece, Type 316 stainless steel with integral work surface and drainage trough.
Equipped with vapor-tight T8 fluorescent light fixture and light and blower switches. Two remote-controlled service fixtures can be used with gas, air, vacuum, or cold water. Each fixture has a serrated-tip hose connector. Hoods are factory prepared for up to eight service fixtures and four electrical duplexes. Hand-operated hood and duct washdown systems. Equipped with 27.9 cm (11") O.D., Type 316 stainless steel collar for connection to the exhaust stack.
Supporting work surface isMore Product Information