Magnesium nitrate (≥10.0 ± 0.2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ100
1.05813.0050
1.05813.0050
CA1.05813.0050CS
14828.08
CAD
EM1.05813.0050
CA1.05813.0050
Magnesium nitrate (≥10.0 ± 0.2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Magnesium nitrate solutions
Magnesium matrix modifier for graphite furnace AAS.
Formula:
Mg(NO₃)₂ Density: 1.05 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
10377-60-3 UN: 2031 ADR: 8,II |
Specification Test Results
Ag (Silver) | ≤ 0.000002 g/l |
As (Arsenic) | ≤ 0.00001 g/l |
Cd (Cadmium) | ≤ 0.000001 g/l |
Pb (Lead) | ≤ 0.00001 g/l |
Se (Selenium) | ≤ 0.0001 g/l |
Tl (Thallium) | ≤ 0.00001 g/l |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...