Anti-LY76 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]

Supplier: Tonbo Biosciences
20-5921-U025 20-5921-U100
10050-638EA 74.26 CAD
10050-638 10050-640
Anti-LY76 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]
Antibodies
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.

Type: Primary
Antigen: LY76
Clonality: Monoclonal
Clone: TER-119
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR