Anti-FOXP3 Rat Antibody (APC (Allophycocyanin)) [clone: MF23]

Supplier: Tonbo Biosciences
20-0191-U025 20-0191-U100
10140-998EA 178.54 CAD
10140-998 10141-000
Anti-FOXP3 Rat Antibody (APC (Allophycocyanin)) [clone: MF23]
Antibodies
The MF23 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is a member of the forkhead box or winged helix family of transcription factors and is expressed at constitutively high levels in Treg cells. Forced expression of Foxp3 in conventional T cells results in the upregulation of Treg associated molecules such as CD25, CTLA-4, and GITR and is sufficient to impart suppressive functional activity to these cells.

Type: Primary
Antigen: Foxp3
Clonality:
Clone: MF23
Conjugation: APC
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR