7-Aminoactinomycin D (7-AAD)

Supplier: Tonbo Biosciences

13-6993-T200 13-6993-T500
10140-986EA 31.6 CAD
10140-986 10140-988
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)

7-AAD (7-Aminoactinomycin D) is a nucleic acid dye that can be used to exclude nonviable cells from the analysis of flow cytometry data based on light scatter and fluorescence properties.


Excitation Laser: Blue (488 nm), Green (532 nm), Yellow-Green (561 nm)
Emission (nm): 647
Formulation: 10 mM NaH2PO4, 150 mM Na10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2Cl, 0.09% NaN3, pH 7.2
Application: Flow Cytometry


7-AAD is provided in solution and should be stored at 2-8°C and protected from light. Do not freeze.


Caution: 7-AAD is a potential carcinogen. Protect skin and eyes by wearing suitable protective clothing, gloves and eye/face protection.

Formula: C₆₂H₈₇N₁₃O₁₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077295
CAS Number: 7240-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR