Anti-TCRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H57-597]

Supplier: Tonbo Biosciences
35-5961-U025 35-5961-U100 35-5961-U500
10051-036EA 36.34 CAD
10051-036 10051-038 10051-040
Anti-TCRB Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H57-597]
Antibodies
The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma/delta protein chains, known as the g/d TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactionsMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol TCRB
Antigen name T-cell Receptor beta
Environmentally Preferable
Antigen synonyms TCRB|T cell receptor beta|TRB|T cell antigen receptor
Antibody type Primary
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone H57-597
Reactivity Mouse
Host Armenian hamster
Gene ID 21577
Isotype IgG
Flow cytometry Yes
Neutralisation Yes
Storage temperature Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
Tested applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR