Nalgene® Ported Biotainer® Bottle and Carboy Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
2560-0490 2560-0489
10118-324CS 240.16 CAD
10118-324 CA10118-314
Nalgene® Ported Biotainer® Bottle and Carboy Closures, Thermo Scientific
Closures Bottle Closures
Designed to aseptically handle solutions in biopharmaceutical, biotechnology, and laboratory liquid transfer applications, these two- and three-ported closures are designed for use with Nalgene® Biotainer® bottles and carboys. Featuring Animal Derived Component-Free (ADCF) components, including seals that are sonically welded to the cap and leakproof closures, the security of high value contents are guaranteed during fluid transfer.
Closures fit 1L to 20L Biotainer® bottles and carboys in PC and PETG.