Polycarbonate Shaker Flasks, TriForest Enterprises
Supplier: TRIFOREST ENTERPRISES INC
FPC3000S FBC3000S FPC1000S FBC1000S FPC2000S FBC2000S FPC0125S FBC0125S FPC0250S FBC0250S FPC0500S FBC0500S
10124-400BX 615.87 CAD
10124-400 10124-402 10124-468 10124-470 10124-472 10124-474 10128-916 10128-918 10128-920 10128-922 10128-924 10128-926
Polycarbonate Shaker Flasks, TriForest Enterprises
Flasks Erlenmeyer Flasks
TriForest shaker flasks are manufactured with virgin, optically clear, non leaching LEXAN® resin that retains clarity after gamma radiation.
  • Certified pyrogen free and DNase/RNase free
  • Flasks fit all standard shaking incubator clamps and can be placed on any platform
  • Autoclavable
Comes standard with a convenient vented DuoCap. Simply remove the dustguard cover to allow sterile exchange, and then replace the cover when ready. There is no hassle or risk associated with switching caps. The TriForest vented DuoCap is two caps in one. The cap isMore Product Information