Titrisol® pH Buffers, MilliporeSigma
Supplier: MilliporeSigma
1.09880.0001
1.09884.0001
1.09886.0001
1.09887.0001
1.09889.0001
1.09890.0001
1.09893.0001
CA1.09880.0001CS
3713
CAD
CA1.09880.0001
CA1.09884.0001
CA1.09886.0001
CA1.09887.0001
CA1.09889.0001
CA1.09890.0001
CA1.09893.0001
Titrisol® pH Buffers, MilliporeSigma
Buffers
pH Calibration Buffers
- Traceable to secondary reference material (SRM) of NIST and PTB
- Buffer concentrate for the preparation of 500 mL of buffer solution
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...