BEE-SAFE™ Nitrile Cleanroom Gloves, High-Tech Conversions
Supplier: High-Tech Conversions
GLH-CRN-12-SM GLH-CRN-12-MD GLH-CRN-12-LG GLH-CRN-12-XL
10142-566BG 77.2 CAD
10142-566 10142-568 10142-570 10142-572
BEE-SAFE™ Nitrile Cleanroom Gloves, High-Tech Conversions
Gloves
Cleanroom gloves are vital to maintaining the quality of your critical environment. Your hands must be protected adequately when exposed to chemicals, microbes or other hazardous elements. Each BEE-SAFE glove offers its own advantage for various applications and industries.
  • 100% synthetic nitrile polymer
  • Latex-free, textured
  • Suitable for cleanroom Class 100 and 1000 (ISO Class 5)
  • Ambidextrous, beaded cuff
  • Packaged in cleanroom compatible bags
  • Manufactured, packaged and tested in ISO 9001 facility
  • Puncture and tear resistant
Ordering information: More Product Information