Septa for GL Caps, Silicone and Silicone/PTFE, Ace Glass Incorporated
Supplier: Ace Glass
7625-01 7625-02 7625-03 7625-04 7625-05 7625-06 7625-07 7625-08 7625-09 7625-10
10099-036PK 35.27 CAD
10099-036 10099-106 10099-108 10099-110 10099-112 10099-404 10099-406 10099-408 10099-410 10099-412
Septa for GL Caps, Silicone and Silicone/PTFE, Ace Glass Incorporated
Septa
These septa are designed to fit the DURAN® open-top cap product family.
Available in both silicone and PTFE-faced silicone for good chemical resistance. Caps can withstand 130 °C (266 °F) vapor or 200 °C (392 °F) dry heat.
Order Now
SPECIFICATIONS
Environmentally Preferable