Pierceable Plug Stopper - For 16 mm Tubes, Globe Scientific
Supplier: Globe Scientific
113151B 113151G 113151R 113151W 113151Y
10142-476PK 451.31 CAD
10142-476 10142-478 10142-480 10142-482 10142-484
Pierceable Plug Stopper - For 16 mm Tubes, Globe Scientific
Stoppers Rubber Stoppers
These pierceable plug stoppers are intended for recapping vacuum blood tubes and test tubes.
  • Thermoplastic elastomer (TPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
The caps are produced from thermoplastic elastomer (TPE) which can easily be pierced with a syringe.
Ordering information: Stoppers are packaged 1000 per bag.