Avanti™ JXN Series High Performance Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
B38619 B38621
10119-926EA 91537.84 CAD
10119-926 10210-212
Avanti™ JXN Series High Performance Centrifuges, Beckman Coulter®
Centrifuges Floor-Standing Centrifuges
The Avanti™ JXN Series of High Performance Centrifuges offer the functionality, design level, performance, and versatility to meet any laboratory's needs.
  • Foot pedal for hands-free operation
  • Remote monitoring and control through Molecule App
  • Friction Reduction System (FRS)
  • Dynamic Rotor Inertia Check (DRIC)
  • BioSafe options
The Avanti™ JXN Series features a low work-surface height, a door that swings up and out of the way for easy rotor chamber access, and large digital readouts
Additional standard features include a high-torque drive that shortens cycle times, BioSafe options, and more. With password protection, e-signature which permits users to electronically add a signature and a note (if desired), numerous stored protocols, and networking across multiple instruments, data management options are ideal for GMP environment or shared laboratories. The Avanti™ JXN Series offers ergonomic design, intuitive interface, remote control and monitoring, safety and sustainability, easy-to-use data management, and versatility across applications.
The intuitive interface, with a 15" LCD screen, provides a view of set and actual run conditions simultaneously and rotor selection by name. The MobileFuge app allows users to access data and user management and remote monitoring and control via networks and mobile devices. Remote control and monitoring enables users to set and monitor run parameters remotely with mobile devices and receive alerts and diagnostics remotely.
Ordering information: Rotors and optional accessories sold separately. For high performance centrifuge bottles and tubes, contact your Avantor sales representative.