Isopropanol, Contec®
Supplier: Contec
SB1287030 SB327030 SB128100 SBT167030
89209-304CS 472.56 CAD
89209-304 10120-202 CA10591-354 CA10120-204 77574-786
Isopropanol, Contec®
Cleaning Agents Detergents and Surface Cleansers
Contec® Isopropanol contains USP grade isopropanol (IPA) and USP grade purified water. Filtered to 0.2 μm, bottles are double-bagged to assure product quality. Each bottle has a lot number code and expiration date for easy record keeping.
  • Contains USP grade isopropanol which evaporates and leaves extremely low residues
  • Filtered through 0.2 μm in-line filter to eliminate the need for in-house filtration
  • Meets USP grade purified water
A convenient trigger sprayer with settings for stream or mist spraying is included with the 32 oz. bottles. The streamMore Product Information