Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HI30]

Supplier: Tonbo Biosciences
35-0459-T025 35-0459-T100 35-0459-T500
10050-932EA 50.56 CAD
10050-932 10050-934 10208-676
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HI30]
Antibodies
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PTPRC PTPRC
Antigen name Protein Tyrosine Phosphatase Receptor Type C Protein Tyrosine Phosphatase Receptor Type C
Environmentally Preferable
Antigen synonyms CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate) FITC (Fluorescein Isothiocyanate)
Clone HI30 HI30
Reactivity Chimpanzee,Human Chimpanzee,Human
Host Mouse Mouse
Gene ID 5788 5788
Isotype IgG1 IgG1 kappa
Flow cytometry Yes Yes
Neutralisation Yes
Storage temperature Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze. 2-8°C protected from light
Tested applications Flow Cytometry Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR