Anti-IFNG Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: XMG1.2]

Supplier: Tonbo Biosciences
20-7311-U025 20-7311-U100
10050-662EA 74.26 CAD
10050-662 10050-664
Anti-IFNG Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: XMG1.2]
Antibodies
The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20 kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells, which allow the cytokine to modulate innate immunity, when secreted by NK and NKT cells, and to function in supporting adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).

The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein by flow cytometry. Other formats can be used for quantitative analysis of the secreted protein by ELISA, when paired with an appropriated secondary antibody. This clone has been reported for neutralization of the functional activity of IFN-g in a variety of assays (use format suitable for functional assays).

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: XMG1.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR