Industrial Single-Wall Storage Cabinets, Securall
Supplier: Securall Cabinets
SS242 SS272 SS284
14227-662EA 3154.2 CAD
14227-662 14227-664 14227-666
Industrial Single-Wall Storage Cabinets, Securall
Furniture Storage Cabinets
Rugged, all welded, 18-gauge steel constructed storage cabinet will provide years of service in both factory and office environments.
  • Three-point self-latching doors add even more security
  • Shipped fully assembled and ready to use
Double wall cabinet doors feature full length piano hinges and a lockable flush mounted door handle for obstruction free, finger tip operation. Each shelf is easily adjustable and carries a 500 lbs. load capacity.