Vertical Flammable Drum Storage Cabinets, Securall
Supplier: Securall Cabinets
V175 V375 V1500
14227-652EA 7008.3 CAD
14227-652 14227-654 14227-656
Vertical Flammable Drum Storage Cabinets, Securall
Furniture Storage Cabinets
Cabinets for vertical drum storage.
  • Color: yellow
Vertical cabinets have ball bearing rollers for easy placing and removal of 55-gallon drums.
Certifications: FM approved, OSHA and NFPA 30 compliant.