Science Take-Out® Acid Rain and Buffers
Supplier: SCIENCE TAKE-OUT, LLC
107 107U 107R 107-NOTG
470017-736EA 27.1 CAD
470017-736 470017-776 470017-844 470017-737
Science Take-Out® Acid Rain and Buffers
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Learn How Certain Types Of Lake Bottoms Can Act As Buffers.
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Why does acid rain decrease the pH of some lakes and not others? Explore the role of different bottom materials in keeping the pH of lakes constant. Create mini-lakes with different bottom materials and limestone. Add acid rain to your mini-lakes and test the pH. Determine which lake bottom material is a buffer that keeps the pH of the mini-lake waterMore Product Information