Science Take-Out® Yeast Populations
Supplier: SCIENCE TAKE-OUT, LLC
123 123U 123R 123-NOTG
470017-764EA 31.05 CAD
470017-764 470017-804 470017-864 470017-765
Science Take-Out® Yeast Populations
Educational Materials Biology Educational Materials Microbiology Learning Activities
What Environmental Conditions Can Affect Organism Growth?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Test samples from a simulated yeast population to determine yeast population size, the pH of the environment, and the amount of sugar present. Graph your results and compare them with results from another researcher. This complete experiment setup contains all materials and does not require lab equipment.