ThermoFinnigan AI3000 and AS 3000 Syringes, Hamilton
Supplier: Hamilton
204000 204001 204051 204052
10118-418EA 259.25 CAD
10118-418 10118-420 10118-422 10118-424
ThermoFinnigan AI3000 and AS 3000 Syringes, Hamilton
Syringes Syringes for Chromatography
Hamilton offers two quality syringe options for the ThermoFinnigan HS 250, HS 500, and HS 850, one quality syringe option for the AS 800 and AS 2000, and four quality syringe options for the AI 3000 and AS 3000 GC Autosamplers.
The Thermo Scientific 3000 Series II of liquid sampling systems for gas chromatography represent the next evolution in GC liquid sampling automation. The 3000 Series II incorporate advanced micro-step motor technology, ensuring excellent precision and fine control of the sampling turret movements. This version complies with the latest restrictions on hazardous substances in electrical and electronic equipment.
Certifications: UL Listed. CE compliant.
Ordering information: Accessories not included.