Science Take-Out® Enzymes And Lactose Intolerance
Supplier: SCIENCE TAKE-OUT, LLC
119 119U 119R 119-NOTG
470017-756EA 38.4 CAD
470017-756 470017-796 470017-858 470017-757
Science Take-Out® Enzymes And Lactose Intolerance
Educational Materials Biology Educational Materials Cell Biology Learning Activities
How Do Enzymes Help People With Lactose Intolerance?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
How does the dietary supplement LACTAID work to help people with lactose intolerance? Model the action of the enzyme lactase. Conduct tests to determine whether LACTAID, an enzyme supplement, digests lactose. Design and conduct an experiment to determine if acid interferes with the action of the enzymes in LACTAID. This complete experiment setup contains all materials and does not require lab equipment.