D-Glucoronic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

M-Clarity™ quality level = MQ200

8.43961.0010 8.43961.0025
CA8.43961.0010EA 165 CAD
CA8.43961.0010 CA8.43961.0025
D-Glucoronic acid for synthesis, Sigma-Aldrich®
D-Glucoronic acid
Formula: C₆H₁₀O₇
MDL Number: MFCD00166981
CAS Number: 6556-12-3

Order Now

Specification Test Results

Solubility (20°C) Soluble
Melting Point 159 - 161 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR