2-Iodopropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.43955.0250 8.43955.1000
CA8.43955.0250EA 293 CAD
CA8.43955.0250 CA8.43955.1000
2-Iodopropane for synthesis, Sigma-Aldrich®
2-Iodopropane
Formula: C₃H₇I
MDL Number: MFCD00001071
CAS Number: 75-30-9
UN: 2392
ADR: 3,III

Order Now

Specification Test Results

Solubility 1.4 g/l (20°C)
Melting Point -90 °C
Flash Point 42 °C
Density 1.703 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR