Adipoyl dichloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.43951.0050 8.43951.0250
CA8.43951.0050CS 1510 CAD
CA8.43951.0050 CA8.43951.0250
Adipoyl dichloride for synthesis, Sigma-Aldrich®
Adipoyl dichloride
Formula: C₆H₈Cl₂O₂
MDL Number: MFCD00000759
CAS Number: 111-50-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.259 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR