Cyclopentyl methyl ether (CPME) stabilised for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.43949.0250 8.43949.1000
CA8.43949.0250CS 804 CAD
CA8.43949.0250 CA8.43949.1000
Cyclopentyl methyl ether (CPME) stabilised for synthesis, Sigma-Aldrich®
Cyclopentyl methyl ether (CPME)
Formula: C₆H₁₂O
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Melting Point -140 °C
Flash Point 59.9 hPa (25 °C)
Refractive Index -1 °C
Density 0.86 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR