2-(Propyloxy)ethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethylene glycol monopropyl ether, 2-Propoxyethanol, Propyl glycol

M-Clarity™ quality level = MQ200

8.43947.0250 8.43947.1000
CA8.43947.0250CS 244 CAD
CA8.43947.0250 CA8.43947.1000
2-(Propyloxy)ethanol for synthesis, Sigma-Aldrich®
2-(Propyloxy)ethanol
Formula: C₅H₁₂O₂
MDL Number: MFCD00072645
CAS Number: 2807-30-9
UN: 1986
ADR: 3 (6.1),III

Order Now

Specification Test Results

Melting Point -75 °C
Vapor Pressure 2.1 hPa (20 °C)
Flash Point 48 °C
Density 0.912 g/cm3 (20 °C)
Ignition Point 230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR