Ethyl-3-methoxypropionate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 3-Methoxypropionic acid ethyl ester

M-Clarity™ quality level = MQ200

8.43946.0250 8.43946.1000
CA8.43946.0250CS 526 CAD
CA8.43946.0250 CA8.43946.1000
Ethyl-3-methoxypropionate for synthesis, Sigma-Aldrich®
Ethyl 3-methoxypropionate
Formula: C₆H₁₂O₃
MDL Number: MFCD00143172
CAS Number: 10606-42-5
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Solubility 52 g/l (20°C)
Boiling Point 166 °C (1013 hPa)
Vapor Pressure 2.0 hPa (25 °C)
Flash Point 58 °C
Density 0.93 g/cm3 (20 °C)
Ignition Point 377°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR