Isobutyl stearate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.43070.0100 8.43070.1000 8.43070.2500
CA8.43070.0100CS 446 CAD
CA8.43070.0100 CA8.43070.1000 CA8.43070.2500
Isobutyl stearate for synthesis, Sigma-Aldrich®
Isobutyl stearate
Formula: C₂₂H₄₄O₂
CAS Number: 646-13-9

Order Now

Specification Test Results

Melting Point 28.9 °C
Density 0.85 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR