2-Mercapto-2-thiazoline for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.21111.0100 8.21111.0500 8.21111.9010
CA8.21111.0100CS 1027 CAD
CA8.21111.0100 CA8.21111.0500 CA8.21111.9010
2-Mercapto-2-thiazoline for synthesis, Sigma-Aldrich®
2-Mercapto-2-thiazoline
Formula: C₃H₅NS₂
Melting Pt: 101…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00126013
CAS Number: 96-53-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20°C) inSoluble
Melting Point 101 - 106 °C
pH 6 - 7 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR