1-Dodecylpyridinium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20546.0005 8.20546.0250 8.20546.1000 8.20546.9025
CA8.20546.0005EA 77 CAD
CA8.20546.0005 CA8.20546.0250 CA8.20546.1000 CA8.20546.9025
1-Dodecylpyridinium chloride for synthesis, Sigma-Aldrich®
1-Dodecylpyridinium chloride
Formula: C₁₇H₃₀ClN
Melting Pt: 87…89 °C
Density: 1.04 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011987
CAS Number: 104-74-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 600 g/l (20°C)
Melting Point 87 - 89 °C
Flash Point 160 °C
Density 1.04 g/cm3 (20 °C)
pH 4 (10 g/l, H2O, 20 °C)
Ignition Point 270°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR