2,3-Butanediol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.18801.0005
8.18801.0250
8.18801.1000
8.18801.9010
CA8.18801.0005EA
41
CAD
CA8.18801.0005
CA8.18801.0250
CA8.18801.1000
CA8.18801.9010
2,3-Butanediol for synthesis, Sigma-Aldrich®
2,3-Butanediol
Mixture of meso- D- and L-form
Formula:
C₄H₁₀O₂ Boiling Pt: 184 °C (1013 hPa) Melting Pt: 18…20 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004523 CAS Number: 513-85-9 EINECS: 208-173-6 Merck Index: 12,01602 |
Specification Test Results
Solubility | 1000 g/l (20°C) |
Melting Point | 18 - 20 °C |
Boiling Point | 180 - 182 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 96 °C DIN 51758 |
Refractive Index | 1.4348 (20 °C, 589 nm) |
Explosion Limit | 3.1 - 11.4 %(V) |
Density | 1.00 g/cm3 (20 °C) |
pH | 7 (H2O, 20 °C) Aqueous solution |
Ignition Point | 395°C DIN 51794 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...