Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Dimethylpropyleneurea, Dimethyl propylene urea, 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 1,3-Dimethyltetrahydropyrimidin-2(1H)-one, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, N,N'-Dimethyl propylene urea

M-Clarity™ quality level = MQ200

8.18214.0100 8.18214.0500 8.18214.9025
CA8.18214.0100CS 1693 CAD
CA8.18214.0100 CA8.18214.0500 CA8.18214.9025
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Tetrahydro-1,3-dimethyl-1H-pyrimidin-2-one
Formula: C₆H₁₂N₂O
MW: 128.17 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.064 g/cm3 (20 °C)
Flash Pt: 121
Storage Temperature: Ambient
MDL Number: MFCD00006550
CAS Number: 7226-23-5

Order Now

Specification Test Results

Solubility (20°C) Soluble
Melting Point -24 °C
Boiling Point 247 °C (1013 hPa)
Vapor Pressure 0.5 hPa (57 °C)
Flash Point 121 °C
Explosion Limit 1.6 - 9.3 %(V)
Density 1.064 g/cm3 (20 °C)
pH 11 (100 g/l, H2O, 20 °C)
Ignition Point 255°C
Synonyms Dimethylpropyleneurea, DMPU
Hill Formula C6H12N2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR