Phthalimide ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07303.0100 8.07303.0500 8.07303.1000 8.07303.9050
CA8.07303.0100EA 42 CAD
CA8.07303.0100 CA8.07303.0500 CA8.07303.1000 CA8.07303.9050
Phthalimide ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Phthalimide
Formula: C₈H₅NO₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 234…238 °C (subl.)
Density: 1.21 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005881
CAS Number: 85-41-6

Order Now

Specification Test Results

Solubility 0.036 g/l (25°C)
Melting Point 234 - 238 °C (sublimed)
Boiling Point 310 °C (1013 hPa)
Flash Point 150 °C
Density 1.21 g/cm3 (20 °C)
pH 3.8 (0.6 g/l, H2O)
Ignition Point 500°C
Synonyms Phthalic acid imide
Hill Formula C8H5NO2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR